
1,2-Diphenylethylamine ≥97.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:25611-78-3
:
Formula:C₁₄H₁₅N
MDL Number:MFCD00008063
:197.28 g/mol
EINECS:247-126-4
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 97.0 %
- Purity (Neutralization titration):min. 97.0 %