Trityl chloride ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:76-83-5
:
Formula:C₁₉H₁₅Cl
MDL Number:MFCD00000813
:278.78 g/mol
Boiling Pt:310°C
Melting Pt:109…112 °C
Density:1,25 g/cm³ (20 °C)
Storage Temperature:Refrigerator
EINECS:200-986-4
UN:3261
ADR:8,III
:GHS05
Specification Test Results
- Appearance:White to Light yellow powder to crystal
- Purity (Argentmetric Titration):min. 98.0 %
- Melting point:110.0 to 115.0 deg-C