n-Hexylbenzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1077-16-3
:
Formula:C₁₂H₁₈
MDL Number:MFCD00009526
:162.27 g/mol
Boiling Pt:226°C
Melting Pt:–61 °C
Density:0,8666 g/cm³ (20 °C)
Flash Pt:83°C
Storage Temperature:Ambient
EINECS:214-070-7
UN:3082
ADR:9,III
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %