
2-Fluoro-5-nitrotoluene ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:455-88-9
:
Formula:C₇H₆FNO₂
MDL Number:MFCD00007284
:155.13 g/mol
Boiling Pt:99°C
Melting Pt:40 °C
Storage Temperature:Refrigerator
EINECS:207-251-7
ADR:6.1,III
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 97.0 %
- Solubility in Methanol:almost transparency
- Melting point:38.0 to 41.0 deg-C