2-Hydroxy-m-toluic acid ≥98.0% (by GC, titration analysis)
: TCI
:83-40-9
:
:C₈H₈O₃
:MFCD00002448
:14,02581
:293 °C (1013 hPa)
:163…165 °C
:152.15 g/mol
:Ambient
:201-473-8
:GHS05
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:165.0 to 169.0 deg-C