
α-Bromo-2-fluorotoulene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:446-48-0
:
Formula:C₇H₆BrF
MDL Number:MFCD00000324
:189.03 g/mol
Boiling Pt:115°C
Density:1,567 g/cm³ (20 °C)
Flash Pt:82°C
EINECS:207-169-1
UN:3265
ADR:8,II
:GHS05
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %