
(R)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetic acid ≥98.0% (by GC, titration analysis) optical resolving
: TCI
:20445-31-2
:
:C₁₀H₉F₃O₃
:MFCD00004184
:234.17 g/mol
:243-829-5
:GHS07
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Optical purity (GC):min. 99.0 ee%
- Purity (Neutralization titration):min. 98.0 %
- Specific rotation [a]20/D:+70.0 to +75.0 deg (C=2, Methanol)