
2-Hydroxyphenylboronic acid pinacol ester ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:269409-97-4
:
Formula:C₁₂H₁₇BO₃
MDL Number:MFCD02093754
:220.08 g/mol
:GHS07
Specification Test Results
- Appearance:Colorless to Yellow clear liquid
- Purity (GC):min. 97.0 %