
Biphenyl-2,2'-dicarboxylic anhydride ≥98.0% (by HPLC, titration analysis)
: TCI
:6050-13-1
:
:C₁₄H₈O₃
:MFCD00004993
:224.22 g/mol
:227-950-0
:GHS07
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Morpholine Method):min. 98.0 %
- Melting point:224.0 to 230.0 deg-C
- Solubility in hot Toluene:almost transparency