

3,4,5-Trifluorophenol ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:99627-05-1
:
Formula:C₆H₃F₃O
MDL Number:MFCD00042427
:148.08 g/mol
Density:1,629 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:436-120-9
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:55.0 to 59.0 deg-C