Order Entry
Finland
ContactUsLinkComponent
2,4,6-Trichloroanisole ≥98.0% (by GC)
2,4,6-Trichloroanisole ≥98.0% (by GC)
Supplier:  TCI
Certificates
undefined
2,4,6-Trichloroanisole ≥98.0% (by GC)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    59.0 to 62.0 deg-C

Specification Test Results