Order Entry
Finland
ContactUsLinkComponent
 
 
 
 
1,2-Dichloro-3-nitrobenzene ≥99.0% (by GC)
 :  TCI
 
undefined
 
 
 
1,2-Dichloro-3-nitrobenzene ≥99.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    60.0 to 63.0 deg-C
  • Solubility in EtOH:
    almost transparency