
2,3-Dichlorobenzaldehyde ≥98.0% (by GC)
: TCI
:6334-18-5
:
:C₇H₄Cl₂O
:MFCD00010127
:175.01 g/mol
:105°C
:64 °C
:1,33 g/cm³ (100 °C)
:Ambient
:228-711-3
:1759
:8,III
:GHS05
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:61.0 to 64.0 deg-C