
4-Methoxysalicylic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:2237-36-7
:
Formula:C₈H₈O₄
MDL Number:MFCD00002450
:168.15 g/mol
Boiling Pt:327 °C (1013 hPa)
Melting Pt:191 °C
EINECS:218-801-0
:GHS07
Specification Test Results
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:158.0 to 162.0 deg-C