
4-Hydroxy-3-nitrobenzoic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:616-82-0
:
Formula:C₇H₅NO₅
MDL Number:MFCD00007119
:183.12 g/mol
Melting Pt:182…185 °C
Density:1 g/cm³ (20 °C)
EINECS:210-494-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to Brown powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Neutralization titration):min. 98.0 %
- Melting point:182.0 to 185.0 deg-C
- Solubility in Methanol:very faint turbidity