
Ethyl 4-hydroxybenzoate ≥99.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:120-47-8
:
Formula:C₉H₁₀O₃
MDL Number:MFCD00002353
Merck Index:14,03837
Boiling Pt:178°C
Melting Pt:116…118 °C
Density:1,078 g/cm³ (131 °C)
:166.18 g/mol
EINECS:204-399-4
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 99.0 %
- Purity (Neutralization titration):min. 99.0 %
- Melting point:115.0 to 118.0 deg-C
- Solubility in Methanol:almost transparency
Specification Test Results
Frequently Bought Together