Order Entry
Finland
ContactUsLinkComponent
 
 
2-Nitrobenzonitrile ≥98.0% (by GC)
  TCIAN0171-5G
 :  TCI
 :  
undefined
 
2-Nitrobenzonitrile ≥98.0% (by GC)
  TCIAN0171-5G
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Appearance:
    Light yellow to Yellow powder to crystaline
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    106.0 to 111.0 deg-C

 

 

 :
 :148.12 g/mol
 :165°C
 :107…109 °C
 :Ambient
 :210-301-0
 :3439
 :6.1,III
 :GHS06
 :612-24-8
 :C₇H₄N₂O₂
 :MFCD00007044