
1-Bromo-4-(trifluoromethoxy)benzene ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:407-14-7
:
Formula:C₇H₄BrF₃O
MDL Number:MFCD00040834
:241.01 g/mol
Boiling Pt:80°C
Density:1,62 g/cm³ (20 °C)
Flash Pt:67°C
Storage Temperature:Ambient
EINECS:206-979-2
ADR:9,
:GHS07
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
Frequently Bought Together