Order Entry
Finland
ContactUsLinkComponent
1-Bromo-3-fluoro-4-iodobenzene ≥99.0% (by GC)
Irritant
1-Bromo-3-fluoro-4-iodobenzene ≥99.0% (by GC)
Catalog # TCIAB1947-25G
Supplier:  TCI
CAS Number:  
undefined
Irritant
1-Bromo-3-fluoro-4-iodobenzene ≥99.0% (by GC)
Catalog # TCIAB1947-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 99.0 %
  • Melting point:
    47.0 to 50.0 deg-C
  • Solubility in Methanol:
    within almost transparency

Specification Test Results

About this item

 :
 :300.9 g/mol
Boiling Pt:241 °C (1013 hPa)
Melting Pt:46…49 °C
 :GHS07
CAS Number:105931-73-5
Formula:C₆H₃BrFI
MDL Number:MFCD00010608