

1-Bromo-4-fluorobenzene ≥98.0% (by GC)
: TCI
:460-00-4
:
:C₆H₄BrF
:MFCD00000342
:153°C
:–16 °C
:1,4946 g/cm³ (20 °C)
:56°C
:Ambient
:207-300-2
:3,III
:175 g/mol
:1993
:GHS02
:GHS07
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %