3,3-Dimethyl-1-butene ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:558-37-2
:
Formula:C₆H₁₂
MDL Number:MFCD00008853
:84.16 g/mol
Boiling Pt:41°C
Melting Pt:–115 °C
Density:0,6683 g/cm³ (20 °C)
Flash Pt:–28°C
Storage Temperature:Refrigerator
EINECS:209-195-9
UN:3295
ADR:3,II
:GHS02
:GHS07
:GHS08
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %