Order Entry
Finland
ContactUsLinkComponent
 
 
 
2-Methyl-2-pentene ≥95.0% (by GC)
 :  TCI
 
undefined
 
 
2-Methyl-2-pentene ≥95.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %