Order Entry
Finland
ContactUsLinkComponent
 
 
1-Hexadecene ≥99.0% (by GC)
 :  TCI
 
undefined
 
1-Hexadecene ≥99.0% (by GC)
 :  TCI

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 99.0 %