


trans-2-Pentenal ≥95.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1576-87-0
:
Formula:C₅H₈O
MDL Number:MFCD00009615
:84.12 g/mol
Boiling Pt:81°C
Density:0,86 g/cm³ (20 °C)
Flash Pt:22°C
EINECS:216-414-1
UN:1989
ADR:3,II
:GHS02
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to Light yellow to Light orange clear liquid
- Purity (GC):min. 95.0 %