Order Entry
Finland
ContactUsLinkComponent
 
10-Undecenal ≥95.0% (by GC)
  TCIAU0011-25ML
 :  TCI
 :  
undefined
10-Undecenal ≥95.0% (by GC)
  TCIAU0011-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Light yellow clear liquid
  • Purity (GC):
    min. 95.0 %

 

 

 :
 :168.28 g/mol
 :103°C
 :7 °C
 :0,8467 g/cm³ (20 °C)
 :100°C
 :Refrigerator
 :203-973-1
 :112-45-8
 :C₁₁H₂₀O
 :MFCD00007032