Order Entry
Finland
ContactUsLinkComponent
2-Methyl-4-heptanone ≥99.0% (by GC)
Flammable
2-Methyl-4-heptanone ≥99.0% (by GC)
Catalog # TCIAM0694-5ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
2-Methyl-4-heptanone ≥99.0% (by GC)
Catalog # TCIAM0694-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 99.0 %

Specification Test Results

About this item

 :
 :128.21 g/mol
Boiling Pt:155°C
Melting Pt:–59 °C
Density:0,8125 g/cm³ (20 °C)
Flash Pt:44°C
EINECS:210-943-1
UN:1224
ADR:3,III
 :GHS02
CAS Number:626-33-5
Formula:C₈H₁₆O
MDL Number:MFCD00027143