
Potassium (2-methoxyphenyl)trifluoroborate ≥98.0% (by HPLC)
Supplier: TCI
Certificates
About this item
CAS Number:236388-46-8
:
Formula:C₇H₇BF₃KO
MDL Number:MFCD04115756
:214.04 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Elemental analysis (Carbon):38.00 to 40.20 %