

1,2-Ethanedithiol ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:540-63-6
:
Formula:C₂H₆S₂
MDL Number:MFCD00004892
:94.2 g/mol
Merck Index:14,03725
Boiling Pt:146°C
Melting Pt:–41 °C
Density:1,374 g/cm³ (20 °C)
Flash Pt:44°C
Storage Temperature:Ambient
EINECS:208-752-3
UN:3336
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %