
1,6-Hexanediol ≥97.0% (by GC)
: TCI
:629-11-8
:
:C₆H₁₄O₂
:MFCD00002985
:118.18 g/mol
:14,04690
:250 °C (1013 hPa)
:39…42 °C
:1,12 g/cm³ (20 °C)
:211-074-0
:GHS07
:Ambient
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 97.0 %
- Melting point:40.0 to 44.0 deg-C
- Solubility in Water:very faint turbidity