1-Nonanol ≥99.0% (by GC)
: TCI
:143-08-8
:
:H₃C(CH₂)₇CH₂OH
:MFCD00002990
:144.26 g/mol
:14,06679
:214°C
:–7 °C
:0,8419 g/cm³ (20 °C)
:104°C
:Ambient
:205-583-7
:3082
:9,III
:GHS07
:GHS08
:GHS09
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %