Order Entry
Finland
ContactUsLinkComponent
 
 
2,2,3,3,4,4-Hexafluoro-1,5-pentanediol ≥98.0% (by GC)
 :  TCI
 
undefined
 
2,2,3,3,4,4-Hexafluoro-1,5-pentanediol ≥98.0% (by GC)
 :  TCI

 

  • Appearance:
    White to Almost white powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Melting point:
    77.0 to 80.0 deg-C