
6-Chloro-1-hexanol ≥96.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2009-83-8
:
Formula:C₆H₁₃ClO
MDL Number:MFCD00002984
:136.62 g/mol
Boiling Pt:117°C
Density:1,0241 g/cm³ (20 °C)
Flash Pt:98°C
EINECS:217-925-2
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 96.0 %