1-Chlorooctane ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:111-85-3
:
Formula:C₈H₁₇Cl
MDL Number:MFCD00001022
:148.68 g/mol
Boiling Pt:183°C
Melting Pt:–58 °C
Density:0,8719 g/cm³ (20 °C)
Flash Pt:64°C
Storage Temperature:Ambient
EINECS:203-915-5
UN:3082
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %