Acetone ≥99.5% (by GC) for spectrophotometry
Catalog # TCIAA0054-500ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.5 %
- Absorbance (330nm):max. 1.000
- Absorbance (340nm):max. 0.100
- Absorbance (350nm):max. 0.020
Specification Test Results
About this item
:
:58.08 g/mol
Merck Index:14,00066
Boiling Pt:56°C
Melting Pt:–95°C
Density:0,792 g/cm³ (20 °C)
Flash Pt:–20°C (closed cup)
Storage Temperature:Ambient
EINECS:200-662-2
UN:1090
ADR:3,II
REACH:01-2119471330-49
:GHS02
:GHS07
CAS Number:67-64-1
Formula:CH₃COCH₃
MDL Number:MFCD00008765