Order Entry
Finland
ContactUsLinkComponent
 
5-Undecanone ≥96.0% (by GC)
  TCIAU0042-25ML
 :  TCI
 :  
undefined
5-Undecanone ≥96.0% (by GC)
  TCIAU0042-25ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    25 mL
  • Seq:
    2315

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 96.0 %

 

 

 :
 :170.3 g/mol
 :106°C
 :Ambient
 :251-371-2
 :33083-83-9
 :C₁₁H₂₂O
 :MFCD00048899