Order Entry
Finland
ContactUsLinkComponent
1-Ethyl-3-(hydroxymethyl)pyridinium ethyl sulphate
1-Ethyl-3-(hydroxymethyl)pyridinium ethyl sulphate
Catalog # TCIAE0682-5G
Supplier:  TCI
CAS Number:  
undefined
1-Ethyl-3-(hydroxymethyl)pyridinium ethyl sulphate
Catalog # TCIAE0682-5G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Yellow to Orange clear liquid
  • Elemental analysis (Carbon):
    43.40 to 47.80 %
  • Elemental analysis (Nitrogen):
    5.05 to 5.60 %
  • Elemental analysis (Sulfuride):
    11.60 to 12.80 %

Specification Test Results

About this item

 :
 :263.31 g/mol
 :GHS07
CAS Number:1064704-03-5
Formula:C₁₀H₁₇NO₅S
MDL Number:MFCD08276375