

Ethyl heptafluorobutyrate ≥97.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:356-27-4
:
Formula:C₆H₅F₇O₂
MDL Number:MFCD00000434
:242.09 g/mol
Boiling Pt:96°C
Density:1,394 g/cm³ (20 °C)
Flash Pt:14°C
EINECS:206-602-1
UN:3272
ADR:3,II
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 97.0 %