Order Entry
Finland
ContactUsLinkComponent
Apigenin ≥98.0% (by HPLC)
Irritant
Apigenin ≥98.0% (by HPLC)
Catalog # TCIAA1514-100MG
Supplier:  TCI
CAS Number:  
undefined
Irritant
Apigenin ≥98.0% (by HPLC)
Catalog # TCIAA1514-100MG
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%

Specification Test Results

About this item

 :
 :270.24 g/mol
Merck Index:14,00730
EINECS:208-292-3
 :GHS07
CAS Number:520-36-5
Formula:C₁₅H₁₀O₅
MDL Number:MFCD00006831