
α'-Chloro-α,α,α-trifluoro-o-xylene ≥98.0% (by GC)
: TCI
:21742-00-7
:
:C₈H₆ClF₃
:MFCD00009920
:194.58 g/mol
:103 - 110 °C (25 -rr)
:1,3079 g/cm³ (20 °C)
:70°C
:244-560-6
:3265
:8,II
:GHS05
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %