
4-Bromo-3-nitrobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:6319-40-0
:
Formula:C₇H₄BrNO₄
MDL Number:MFCD00272137
:246.02 g/mol
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Purity (Neutralization titration):min. 98.0 %
- Melting point:202.0 to 206.0 deg-C