
Potassium oleate ≥98.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:143-18-0
:
Formula:C₁₈H₃₃KO₂
MDL Number:MFCD00064243
:320.56 g/mol
Merck Index:14,07650
Melting Pt:235…240 °C
EINECS:205-590-5
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (Nonaqueous Titration):min. 98.0 %