di-Sodium β-glycerophosphate pentahydrate ≥95.0% (by titrimetric analysis)
Supplier: TCI
Certificates
About this item
CAS Number:13408-09-8
:819-83-0
Formula:C₃H₇Na₂O₆P·5H₂O
MDL Number:MFCD00002135
:306.11 g/mol
Melting Pt:102°C
EINECS:212-464-3
Specification Test Results
- Appearance:White powder to crystal
- Purity (Neutralization titration):min. 95.0 % (calcd.on anh.substance)
- Solubility in Water:almost transparency
- Water:27.0 to 32.0 %
- Disodium alpha-glycerophosphate:max. 4.0 %
Specification Test Results
Frequently Bought Together