Light green SF yellowish
Catalog # TCIAL0054-25G
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:25 g
- Seq:3308
Specifications
Specification Test Results
- Appearance:Orange to Brown to Dark purple powder to crystal
- Lambda max.:631.0 to 635.0 nm (H2O)
- Absorbance (E1%1cm):min. 700 (H2O, 631.0 to 635.0 nm)
Specification Test Results
About this item
:
:792.86 g/mol
Merck Index:14,05485
Melting Pt:288…289 °C
Storage Temperature:Ambient
EINECS:225-906-5
CAS Number:5141-20-8
Formula:C₃₇H₃₄N₂Na₂O₉S₃
MDL Number:MFCD00012121