


Dicyclohexylcarbodiimide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:538-75-0
:
Formula:C₁₃H₂₂N₂
MDL Number:MFCD00011659
Merck Index:14,03096
Boiling Pt:148 - 152 °C (15 hPa)
Melting Pt:35…36 °C
Density:0,95 g/cm³ (40 °C)
Flash Pt:113 °C
UN:2811
ADR:6.1,II
:206.33 g/mol
EINECS:208-704-1
:GHS05
:GHS06
:GHS08
Specification Test Results
- Appearance:White to Almost white powder to lump
- Purity (GC):min. 98.0 %