

Bis(trimethylsilyl)carbodiimide ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:1000-70-0
:
Formula:C₇H₁₈N₂Si₂
MDL Number:MFCD00051538
:186.4 g/mol
Boiling Pt:164°C
Melting Pt:–23 °C
Density:0,8211 g/cm³ (25 °C)
Flash Pt:41°C
EINECS:213-673-2
UN:1993
ADR:3,III
:GHS02
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 %