
2-Amino-5-nitrobenzonitrile ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:17420-30-3
:
Formula:C₇H₅N₃O₂
MDL Number:MFCD00007362
:163.14 g/mol
Melting Pt:210 °C
EINECS:241-446-8
:GHS07
Specification Test Results
- Appearance:Light yellow to Brown powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:209.0 to 212.0 deg-C
- Solubility in Pyridine:almost transparency