Order Entry
Finland
ContactUsLinkComponent
(S)-(-)-1,2-Epoxypropane ≥98.0% (by GC)
Flammable
Toxic
Health
(S)-(-)-1,2-Epoxypropane ≥98.0% (by GC)
Catalog # TCIAP0951-1ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Toxic
Health
(S)-(-)-1,2-Epoxypropane ≥98.0% (by GC)
Catalog # TCIAP0951-1ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 98.0 %
  • Optical purity (GC):
    min. 98.0 ee%

Specification Test Results

About this item

 :
 :58.08 g/mol
Boiling Pt:34°C
Melting Pt:–112 °C
Density:0,8268 g/cm³ (20 °C)
Flash Pt:-37°C
Flash Pt:–37°C
Storage Temperature:Refrigerator
EINECS:240-241-0
UN:1280
ADR:3,I
 :GHS02
 :GHS06
 :GHS08
CAS Number:16088-62-3
Formula:C₃H₆O
MDL Number:MFCD00064312