
5-Bromo-2-hydroxybenzyl alcohol ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:2316-64-5
:
Formula:C₇H₇BrO₂
MDL Number:MFCD00004618
:203.04
Merck Index:12,01460
Boiling Pt:302 °C (1013 hPa)
Melting Pt:113 °C
EINECS:219-026-0
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:105.0 to 110.0 deg-C