
Pyridinium chloride ≥98.0% (by HPLC, titration analysis)
Supplier: TCI
Certificates
About this item
CAS Number:628-13-7
:
Formula:C₅H₆ClN
MDL Number:MFCD00012802
:115.56 g/mol
Boiling Pt:222,5 °C (1013 hPa)
Melting Pt:144 °C
Density:1,34 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:211-027-4
:GHS07
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (Nonaqueous Titration):min. 98.0 %