
Methyl cyanoacetate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:105-34-0
:
Formula:C₄H₅NO₂
MDL Number:MFCD00001939
:99.09 g/mol
Boiling Pt:205°C
Melting Pt:–22.5 °C
Density:1,13 g/cm³ (20 °C)
Flash Pt:105°C
EINECS:203-288-8
:GHS07
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %